Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML3187

Sigma-Aldrich

WIN 18446

≥98% (HPLC)

Synonym(s):

Bis-diamine, Bisdiamine, Fertilysin, N,N′-1,8-Octanediylbis[2,2-dichloro-acetamide; N,N′-Bis(dischloroacetyl)-1,8-octamethylenediamine, NSC 59354, NSC59354, WIN 18,446

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20Cl4N2O2
CAS Number:
Molecular Weight:
366.11
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(NCCCCCCCCNC(C(Cl)Cl)=O)C(Cl)Cl

InChI

1S/C12H20Cl4N2O2/c13-9(14)11(19)17-7-5-3-1-2-4-6-8-18-12(20)10(15)16/h9-10H,1-8H2,(H,17,19)(H,18,20)

InChI key

FAOMZVDZARKPFJ-UHFFFAOYSA-N

Biochem/physiol Actions

WIN 18,446 is an orally available bisdichloroacetyldiamine that potently inhibits aldehyde dehydrogenase family 1 (ALDH1) members-catalyzed all-trans retinoic acid (atRA) production (IC50 = 88 nM/human testicular S10 fraction, 102 nM/ALDH1A1, 187 nM/ALDH1A3) via retinal oxidation, efficiently inhibits ALDH1A2 enzymatic activity in a time-dependent manner (kinact = 22.0/h, Ki = 1026 nM). The inhibitory activity of WIN 18,446 against atRA production forms the basis of its in vivo efficacy as a male contraceptive in inhibiting spermatogenesis among mulitple mammalian species and zebrafish.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service