Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML3119

Sigma-Aldrich

SHP1 Inhibitor, TPI-1

≥98% (HPLC)

Synonym(s):

2-(2,5-Dichloro-phenyl)-[1,4]benzoquinone; 2-(2?,5?-Dichlorophenyl)-1,4-benzoquinone, 2-(2,5-Dichlorophenyl)-2,5-cyclohexadiene-1,4-dione, TPI 1, TPI-1, TPI1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6Cl2O2
CAS Number:
Molecular Weight:
253.08
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C=CC(C(C2=C(C=CC(Cl)=C2)Cl)=C1)=O

InChI

1S/C12H6Cl2O2/c13-7-1-3-11(14)9(5-7)10-6-8(15)2-4-12(10)16/h1-6H

InChI key

ZKHFYORNAYYOTM-UHFFFAOYSA-N

Biochem/physiol Actions

TPI-1 is an orally bioavailable, noncytotoxic benzoquinone compound that acts as a potent irreversible inhibitor of SHP-1 (Src homology region 2 domain-containing phosphatase 1) catalytic domain (IC50 ~ 40 nM or 0.01 μg/ml) with selectivity over SHP-2 (IC50 ~ 0.1 μg/ml), MKP1 (IC50 > 1 μg/ml) and CD45. TPI-1 potently induces mouse and human primary IFN-g+ cells both in vitro and in vivo. Effectively inhibits B16 melanoma tumors in mice (1 or 3 mg/kg, s.c., daily) and prolong the survival. TPI-1 is shown to be 50-fold more effective than sodium stibogluconate in cell-based assays and may non-specifically react with cysteine residues.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service