Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2939

Sigma-Aldrich

S2C3 Dibromide

≥98% (HPLC)

Synonym(s):

CYDE-21 dibromide, N,N′-(Decane-1,10-diyl)bis(9-amino-2,3-dihydro-1H-cyclopenta[b]quinolinium) dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H44N4·2Br
Molecular Weight:
668.55
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

NC1=C2C=CC=CC2=[N+](C3=C1CCC3)CCCCCCCCCC[N+]4=C5CCCC5=C(C6=C4C=CC=C6)N

InChI

1S/C34H42N4/c35-33-25-15-7-9-19-29(25)37(31-21-13-17-27(31)33)23-11-5-3-1-2-4-6-12-24-38-30-20-10-8-16-26(30)34(36)28-18-14-22-32(28)38/h7-10,15-16,19-20,35-36H,1-6,11-14,17-18,21-24H2/p+2

InChI key

MEFIQYBUDHNVHT-UHFFFAOYSA-P

Biochem/physiol Actions

HIV-1 fusion inhibitor that specifically binds to the gp41 membrane proximal external region (MPER)
S2C3, an analog of dequalinium, is an HIV-1 fusion inhibitor that specifically binds to the gp41 membrane proximal external region (MPER) of the HIV-1 envelope spike and blocks CD4-induced conformational changes. S2C3 effectively block HIV-1 infection. S2C3 (CYDE-21) is a cationic amphiphilic bolaamphiphile nano-sized vesicles or vesicle-like aggregates suitable to carry antisense oligonucleotides (ASO) into bacterium such as C. difficile.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cationic amphiphilic bolaamphiphile-based delivery of antisense oligonucleotides provides a potentially microbiome sparing treatment for C. difficile
The Journal of Antibiotics, 71 (2018)
HIV-1 fusion inhibitors targeting the membrane-proximal external region of Env spikes
Nature Chemical Biology, 16 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service