Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2857

Sigma-Aldrich

LHVS

≥98% (HPLC)

Synonym(s):

(S)-(E)-3-(4-Morpholine-carbonylleucyl)amino-5-phenyl-1-phenylsulfonyl-1-pentene, Morpholinurea-leucine-homophenylalanine-vinylsulfone-phenyl, Mu-Leu-Hph-VSPh, Mu-Leu-HphVSPh

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$127.00
25 MG
$517.00

$127.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$127.00
25 MG
$517.00

About This Item

Empirical Formula (Hill Notation):
C28H37N3O5S
CAS Number:
Molecular Weight:
527.68
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$127.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N[C@@H](CC(C)C)C(N[C@H](/C=C/S(=O)(C1=CC=CC=C1)=O)CCC2=CC=CC=C2)=O)N3CCOCC3

InChI key

YUMYYTORLYHUFW-MSKIIMLESA-N

Biochem/physiol Actions

Irreversible, potent and selective cathepsin S inhibitor in vitro and in vivo.
LHVS is an irreversible, potent and selective cathepsin S inhibitor (CatS Ki = 5.9 nM; CatO2/L/B Ki = 0.23/0.72/39 μM, Cruzain Ki = 0.22 μM) that targets CatS active site thiol for covalent modification via its vinyl sulfone moiety. LHVS is a useful tool for studyting CatS-mediated responses in cultures (typical dosing ranges: 25 nM to 10μM) and in animals in vivo, including CatS-induced itch in mice (30 mg/kg sc.), morphine-induced antinociceptive tolerance in rats (50 nmol/rat itrathetically) and mechanical hyperalgesia in nerve-injured rats (3-30 mg/kg sc.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service