Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML2817

Sigma-Aldrich

BAY-1797

≥98% (HPLC)

Synonym(s):

N-(4-(3-Chlorophenoxy)-3-sulfamoylphenyl)-2-phenylacetamide, N-[3-(Aminosulfonyl)-4-(3-chlorophenoxy)phenyl]benzeneacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17ClN2O4S
CAS Number:
Molecular Weight:
416.88
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=CC(OC2=C(C=C(NC(CC3=CC=CC=C3)=O)C=C2)S(=O)(N)=O)=C1

Biochem/physiol Actions

BAY-1797 orally available, potent and selective P2X4 antagonist that is suitable for in vivo studies in rodents. BAY-1797 exhibits potent anti-inflammatory and anti-nociceptive effects in mouse CFA inflammatory pain model.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Werner et al.
Journal of medicinal chemistry, 62(24), 11194-11217 (2019-11-21)
The P2X4 receptor is a ligand-gated ion channel that is expressed on a variety of cell types, especially those involved in inflammatory and immune processes. High-throughput screening led to a new class of P2X4 inhibitors with substantial CYP 3A4 induction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service