Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2801

Sigma-Aldrich

Isoforskolin

≥95% (HPLC)

Synonym(s):

6-Acetyl-7-deacetylforskolin, (3R,6S,6aS)-6-(acetyloxy)-3-ethenyldodecahydro-5S,10S,10bS-trihydroxy-3,4aR,7,7,10aR-pentamethyl-1H-naphtho[2,1-b]pyran-1-one, Coleonol B, 6β-Acetoxy-1α,7β,9α-trihydroxy-8,13-epoxy-labd-14-en-11-one, Isoforskolin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O7
CAS Number:
Molecular Weight:
410.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

optical activity

[α]/D -8 to -5°, c = 0.5 in chloroform-d

color

white to beige

solubility

DMF:H2O (9:1): 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CC(=O)O[C@@H]1[C@H](O)[C@@]2(C)O[C@](C)(CC(=O)[C@]2(O)[C@@]3(C)[C@@H](O)CCC(C)(C)C13)C=C

InChI

1S/C22H34O7/c1-8-19(5)11-14(25)22(27)20(6)13(24)9-10-18(3,4)16(20)15(28-12(2)23)17(26)21(22,7)29-19/h8,13,15-17,24,26-27H,1,9-11H2,2-7H3/t13-,15-,16?,17-,19-,20-,21+,22-/m0/s1

InChI key

CLOQVZCSBYBUPB-VDCUXWMXSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Isoforskolin is a naturally occurring diterpene with anti-inflammatory activity. It has been shown to downregulate the expression of proinflammatory compounds. In an experimental mouse model of Lyme Disease induced arthritis isoforskolin downregulated the transcription and expression of TNF-α and IL-6 induced by recombinant Borrelia burgdorferi basic membrane protein A (rBmpA). In a rat asthmatic model isoforskolin down-regulated production of transforming growth factor β1 (TGF-β1) and interleukin-1β (IL-1β) which was accompanied by inhibition of inflammation and airway remodeling.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service