Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2644

Sigma-Aldrich

NCGC00135472

≥98% (HPLC)

Synonym(s):

3,4-Difluoro-N-[3-(3-methylimidazo[1,2-a]pyrimidin-2-yl)phenyl]-benzamide, C2A

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$116.00
25 MG
$377.00

$116.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$116.00
25 MG
$377.00

About This Item

Empirical Formula (Hill Notation):
C20H14F2N4O
CAS Number:
Molecular Weight:
364.35
UNSPSC Code:
12352200
NACRES:
NA.77

$116.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

Fc1c(ccc(c1)C(=O)Nc2cc(ccc2)c3nc4[n](c3C)cccn4)F

InChI

1S/C20H14F2N4O/c1-12-18(25-20-23-8-3-9-26(12)20)13-4-2-5-15(10-13)24-19(27)14-6-7-16(21)17(22)11-14/h2-11H,1H3,(H,24,27)

InChI key

YOVZCQZBMAXLPR-UHFFFAOYSA-N

Biochem/physiol Actions

NCGC00135472 (C2A) is a resolvin D1 (RvD1) mimetic that activates resolving D1 receptor (DRV1/GPR32) and stimulate phagocytosis of serum-treated zymosan. NCGC00135472 enhances macrophage phagocytosis of live E. coli.
resolvin D1 (RvD1) mimetic that activates DRV1/GPR32

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nan Chiang et al.
Cell chemical biology, 26(2), 244-254 (2018-12-18)
Resolution of acute inflammation is governed, in part, by specialized pro-resolving mediators, including lipoxins, resolvins, protectins, and maresins. Among them, resolvin D1 (RvD1) exhibits potent pro-resolving functions via activating human resolvin D1 receptor (DRV1/GPR32). RvD1 is a complex molecule that requires

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service