Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2574

Sigma-Aldrich

Candicidin

≥50% (HPLC)

Synonym(s):

Candicidin, Candeptin, Candimon, Levorin, Vanobid

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$184.00

$184.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$184.00

About This Item

Empirical Formula (Hill Notation):
C59H84N2O18
CAS Number:
Molecular Weight:
1109.30
UNSPSC Code:
51111800
NACRES:
NA.77

$184.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

Streptomyces griseus

form

solid

storage condition

protect from light

concentration

≥50% (HPLC)

solubility

DMSO: 1 mg/mL

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Candicidin is a polyene macrolide antifungal antibiotic which inhibits large number of fungi. 2 Polyene antifungal antibiotics, such as Candicidin, specifically target cell membranes containing sterols. Thus, selectively targets variety of prokaryotic organisms (such as Candida albicans) and not eukaryotic organisms. Candicidin generates cell membrane damage to Candida albicans (C.albicans) by triggering a rapid efflux of K+ ions. Interestingly, it was found that dosage needed for Candicidin to inhibit growth of stationary phase C. albicans cells is 25 μg/ml whereas log-phase C. albicans cells is only 5 μg/mL. In 1953 Kigman and Lewis performed the first in vivo study with mice to reveal the effects of Candicidin. Their study showed that Candicidin exhibits protective effect against C. albicans, Blastomyces dermatitidis and Sporotrichum schenkii but only a partial protective effect against torulosis and histoplasmosis. LD50 for Candicidin oral administration and intraperitoneal administration in mice were 90-400mg/kg and 2.1-7.0 mg/kg respectively. When comparing the activity of Candicidin to other antifungal compounds in vivo protects 100% of mice with C. albicans infection, whereas usage of twice the amount of Nystatin protects only 60% of the mice. Candicidin may undergo a photoisomerization process, yielding all-trans derivatives of Candicidin when exposed to direct UV light.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service