Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2538

Sigma-Aldrich

KL-2

≥98% (HPLC)

Synonym(s):

N-(5-Chloro-2-methylphenyl)-4-(4-fluorophenyl)-2-hydroxy-4-oxo-2-butenamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13ClFNO3
CAS Number:
Molecular Weight:
333.74
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CC=C(Cl)C=C1NC(/C(O)=C/C(C2=CC=C(F)C=C2)=O)=O

Biochem/physiol Actions

Disruptor of SEC (super elongation complex)
KL-2 is a disruptor of the super elongation complex (SEC). KL-1 blocks the interactions of positive transcription elongation factorb (P-TEFb) and the SEC-scaffolding protein AFF4 resulting in decreased Polymerase II (Pol II) activity and production of Pol II-transcribed genes. KL-2 was shown to cause downregulation of heat shock genes, and was non-toxic and active in an animal model of cancer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaiwei Liang et al.
Cell, 175(3), 766-779 (2018-10-20)
The super elongation complex (SEC) is required for robust and productive transcription through release of RNA polymerase II (Pol II) with its P-TEFb module and promoting transcriptional processivity with its ELL2 subunit. Malfunction of SEC contributes to multiple human diseases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service