Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2388

Sigma-Aldrich

SW157765

≥98% (HPLC)

Synonym(s):

3-(2-Furanyl)-N-[2-(4-pyridinyl)-5-benzoxazolyl]-2-propenamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$125.00
25 MG
$502.00

$125.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$125.00
25 MG
$502.00

About This Item

Empirical Formula (Hill Notation):
C19H13N3O3
CAS Number:
Molecular Weight:
331.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$125.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(c2cc3nc([o]c3cc2)c4ccncc4)C(=O)\C=C\c1[o]ccc1

InChI

1S/C19H13N3O3/c23-18(6-4-15-2-1-11-24-15)21-14-3-5-17-16(12-14)22-19(25-17)13-7-9-20-10-8-13/h1-12H,(H,21,23)/b6-4+

InChI key

QSSYRORXXFTVFI-GQCTYLIASA-N

Biochem/physiol Actions

SW157765 is a selective inhibitor of the non-canonical glucose transporter GLUT8 (SLC2A8), which exhibit potent anticancer activity on KRAS/KEAP1 mutant NSCLC cells that selectively depend on GLUT8 for glucose intake to support serine biosynthesis.
selective inhibitor of the non-canonical glucose transporter GLUT8 (SLC2A8)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth A McMillan et al.
Cell, 173(4), 864-878 (2018-04-24)
Diversity in the genetic lesions that cause cancer is extreme. In consequence, a pressing challenge is the development of drugs that target patient-specific disease mechanisms. To address this challenge, we employed a chemistry-first discovery paradigm for de novo identification of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service