Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2324

Sigma-Aldrich

NLX-112 hydrochloride

≥98% (HPLC)

Synonym(s):

(3-Chloro-4-fluorophenyl)[4-fluoro-4-[[[(5-methyl-2-pyridinyl)methyl]amino]methyl]-1-piperidinyl]methanone hydrochloride, Befiradol hydrochloride, F13640 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22ClF2N3O · xHCl
CAS Number:
Molecular Weight:
393.86 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC2(CCN(CC2)C(=O)c3cc(c(cc3)F)Cl)CNCc1ncc(cc1)C

InChI

1S/C20H22ClF2N3O/c1-14-2-4-16(25-11-14)12-24-13-20(23)6-8-26(9-7-20)19(27)15-3-5-18(22)17(21)10-15/h2-5,10-11,24H,6-9,12-13H2,1H3

InChI key

PKZXLMVXBZICTF-UHFFFAOYSA-N

Biochem/physiol Actions

NLX-112 (also known as befiradol or F13640) is a highly selective 5-HT1A receptor full agonist with >1000-fold selectivity compared to other types of receptors. It has been investigated for the treatment of Parkinson′s disease patients who exhibit L-DOPA-induced dyskinesia
Serotonin 5-HT1A receptor full agonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew C McCreary et al.
Neuropharmacology, 105, 651-660 (2016-01-19)
Although l-DOPA alleviates the motor symptoms of Parkinson's disease (PD), it elicits troublesome l-DOPA-induced dyskinesia (LID) in a majority of PD patients after prolonged treatment. This is likely due to conversion of l-DOPA to dopamine as a 'false neurotransmitter' from
Adrian Newman-Tancredi et al.
The Journal of pharmacy and pharmacology, 69(9), 1178-1190 (2017-06-15)
NLX-112 (befiradol, F13640) is a selective serotonin 5-HT1A receptor agonist. Although it has been tested in vivo, little has been reported on its in vitro signal transduction profile. NLX-112 was tested on G-protein activation, inhibition of adenylyl cyclase, ERK1/2 phosphorylation (pERK)
H Iderberg et al.
Experimental neurology, 271, 335-350 (2015-06-04)
L-DOPA is the gold-standard treatment for Parkinson's disease (PD), but induces troublesome dyskinesia after prolonged treatment. This is associated with the 'false neurotransmitter' conversion of L-DOPA to dopamine by serotonin neurons projecting from the raphe to the dorsal striatum. Reducing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service