Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2322

Sigma-Aldrich

Ilaprazole

≥98% (HPLC)

Synonym(s):

2-[[(4-Methoxy-3-methyl-2-pyridinyl)methyl]sulfinyl]-6-(1H-pyrrol-1-yl)-1H-benzimidazole, IY 81149, IY-81149

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$137.50
25 MG
$547.00

$137.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$137.50
25 MG
$547.00

About This Item

Empirical Formula (Hill Notation):
C19H18N4O2S
CAS Number:
Molecular Weight:
366.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$137.50


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[S](=O)(Cc4nccc(c4C)OC)c1[nH]c2c(n1)cc(cc2)[n]3cccc3

InChI

1S/C19H18N4O2S/c1-13-17(20-8-7-18(13)25-2)12-26(24)19-21-15-6-5-14(11-16(15)22-19)23-9-3-4-10-23/h3-11H,12H2,1-2H3,(H,21,22)

InChI key

HRRXCXABAPSOCP-UHFFFAOYSA-N

Biochem/physiol Actions

Ilaprazole is a proton pump inhibitor (H+/K+ ATPase inhibitor) that suppresses acids secretion. Ilaprazole is a potent and selective inhibitor of T-cell-originated protein kinase (TOPK) that suppresses cancer growth.
proton pump inhibitor (H+/K+ ATPase inhibitor)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S Shin et al.
Alimentary pharmacology & therapeutics, 40(5), 548-561 (2014-07-22)
Ilaprazole, a proton pump inhibitor (PPI) currently in clinical use, may provide improved acid suppression vs. other PPIs. To compare the pharmacodynamic and pharmacokinetic profiles of ilaprazole and esomeprazole. A phase 1, randomised, open-label, single-centre, 4-period crossover study was conducted
D Kwon et al.
Arzneimittel-Forschung, 51(3), 204-213 (2001-04-18)
The inhibitory effects of IY-81149 (2-[[(4-methoxy-3-methyl)-2- pyridinyl]methyl-sulfinyl]-5-(1H-pyrol-1-yl)-1H-benzimidazole, CAS 172152-36-2), a newly developed proton pump inhibitor (PPI) on gastric acid secretion were investigated in vitro and in vivo. In rabbit parietal cell preparation, IY-81149 irreversibly inhibited H+/K(+)-ATPase in dose-dependent manner with
Mengzhu Zheng et al.
Oncotarget, 8(24), 39143-39153 (2017-04-08)
T-cell-originated protein kinase (TOPK) is highly and frequently expressed in various cancer tissues and plays an indispensable role in the mitosis of cancer cells, and therefore, it is an important target for drug treatment of tumor. Ilaprazole was identified to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service