Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2305

Sigma-Aldrich

BPK-29

≥98% (HPLC)

Synonym(s):

2-Chloro-N-[hexahydro-1-[4-(4-morpholinyl)benzoyl]-1H-azepin-4-yl]-N-(phenylmethyl)-acetamide, N-Benzyl-2-chloro-N-[1-(4-morpholin-4-yl-benzoyl)-azepan-4-yl]-acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$139.00
25 MG
$450.00

$139.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$139.00
25 MG
$450.00

About This Item

Empirical Formula (Hill Notation):
C26H32ClN3O3
CAS Number:
Molecular Weight:
470.00
UNSPSC Code:
12352200
NACRES:
NA.77

$139.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(CCl)N(C1CCN(C(C2=CC=C(N3CCOCC3)C=C2)=O)CCC1)CC4=CC=CC=C4

InChI

1S/C26H32ClN3O3/c27-19-25(31)30(20-21-5-2-1-3-6-21)24-7-4-13-29(14-12-24)26(32)22-8-10-23(11-9-22)28-15-17-33-18-16-28/h1-3,5-6,8-11,24H,4,7,12-20H2

InChI key

ZKWKNYRAPOSUDO-UHFFFAOYSA-N

Biochem/physiol Actions

BPK-29 is a potent and selective NR0B1 ligand that covalently binds to NR0B1 Cys 274. BPK-29 inhibits the NR0B1-SNW1 interaction. It inhibits the anchorage-independent growth of KEAP1-mutant cancer cells.
BPK-29 is a small molecule inhibitor that blocks the nuclear factor erythroid 2–related factor 2 (NRF2) pathway.[1] It targets the cysteine 274 residue in the nuclear receptor subfamily 0 group B member 1 (NR0B1) and disrupts protein-protein interactions in non-small-cell lung cancer (NSCLC) cell proteomes.[2]
potent and selective NR0B1 ligand that covalently binds to NR0B1 Cys 274

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel Biology and Druggable Targets via Chemoproteomics.
Alan Saghatelian
Biochemistry, 56(50), 6515-6516 (2017-12-08)
Liron Bar-Peled et al.
Cell, 171(3), 696-709 (2017-10-03)
The transcription factor NRF2 is a master regulator of the cellular antioxidant response, and it is often genetically activated in non-small-cell lung cancers (NSCLCs) by, for instance, mutations in the negative regulator KEAP1. While direct pharmacological inhibition of NRF2 has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service