Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML2252

Sigma-Aldrich

TP053

≥98% (HPLC)

Synonym(s):

N-Methyl-6-nitro-2-phenylthieno[2,3-d]pyrimidin-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N4O2S
CAS Number:
Molecular Weight:
286.31
UNSPSC Code:
51111800

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CNC1=NC(C2=CC=CC=C2)=NC3=C1C=C(S3)[N+]([O-])=O

Biochem/physiol Actions

TP053 is a potent thienopyrimidine antitubercular agent that kills both replicating and nonreplicating Mycobacterium tuberculosis. It appears that TP053 is a prodrug, which is activated by mycothiol-dependent reductase Rv2466c.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Albesa-Jové et al.
The Journal of biological chemistry, 290(52), 31077-31089 (2015-11-08)
Rv2466c is a key oxidoreductase that mediates the reductive activation of TP053, a thienopyrimidine derivative that kills replicating and non-replicating Mycobacterium tuberculosis, but whose mode of action remains enigmatic. Rv2466c is a homodimer in which each subunit displays a modular
Leonardo Astolfi Rosado et al.
The Journal of biological chemistry, 292(32), 13097-13110 (2017-06-18)
The Mycobacterium tuberculosis rv2466c gene encodes an oxidoreductase enzyme annotated as DsbA. It has a CPWC active-site motif embedded within its thioredoxin fold domain and mediates the activation of the prodrug TP053, a thienopyrimidine derivative that kills both replicating and
David Albesa-Jové et al.
ACS chemical biology, 9(7), 1567-1575 (2014-06-01)
The emergence of multidrug- and extensively drug-resistant strains of Mycobacterium tuberculosis highlights the need to discover new antitubercular agents. Here we describe the synthesis and characterization of a new series of thienopyrimidine (TP) compounds that kill both replicating and non-replicating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service