Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2251

Sigma-Aldrich

Nanchangmycin

≥98% (HPLC)

Synonym(s):

(2S,4R,6E,8S)-8-[(2S,2′R,4′S,5R,5′S,7R,7′R,8R,9S,9′S,10′R)-9-hydroxy-2,4′,8,10′-tetramethyl-2′-[(2S,3S,5R,6R)-tetrahydro-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyl-2H-pyran-2-yl]-9′-[[(2S,5S,6R)-tetrahydro-5-methoxy-6-methyl-2H-pyran-2-yl]oxy][2,7′-bi-1,6-dioxaspiro[4.5]dec]-7-yl]-2,4,6-trimethyl-5-oxo-6-nonenoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$177.00
25 MG
$715.00

$177.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$177.00
25 MG
$715.00

About This Item

Empirical Formula (Hill Notation):
C47H78O14
CAS Number:
Molecular Weight:
867.11
UNSPSC Code:
12352200
NACRES:
NA.77

$177.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

Streptomyces nanchangensis

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

OC([C@@H](C)C[C@@H](C)C(/C(C)=C/[C@H](C)[C@]1([H])[C@H](C)[C@@H](O)C[C@]2(CC[C@]([C@@]3([H])C[C@H](O[C@@]4([H])O[C@H](C)[C@@H](OC)CC4)[C@@H](C)[C@@]5(O[C@]([C@@]6([H])O[C@@](O)(CO)[C@H](C)C[C@@H]6C)([H])C[C@@H]5C)O3)(C)O2)O1)=O)=O

InChI

1S/C47H78O14/c1-24(40(50)25(2)18-28(5)43(51)52)17-26(3)41-31(8)34(49)22-45(59-41)16-15-44(11,61-45)38-21-36(56-39-14-13-35(54-12)33(10)55-39)32(9)47(58-38)30(7)20-37(57-47)42-27(4)19-29(6)46(53,23-48)60-42/h17,25-39,41-42,48-49,53H,13-16,18-23H2,1-12H3,(H,51,52)/b24-17+

InChI key

FELYAZAWTURXNF-JJIBRWJFSA-N

Biochem/physiol Actions

Nanchangmycin is a broad spectrum polyether antibiotic recently shown to inhibit Zika virus entry. Nanchangmycin was found to potently inhibit entry in three distinct cell types including human brain microvascular endothelial cells (HBMECs) with two different strains of Zika virus. It was also found effective against other viruses such as West Nile, dengue, and chikungunya.
Polyether antibiotic shown to inhibit Zika virus entry

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keiko Rausch et al.
Cell reports, 18(3), 804-815 (2017-01-19)
Zika virus is an emerging arthropod-borne flavivirus for which there are no vaccines or specific therapeutics. We screened a library of 2,000 bioactive compounds for their ability to block Zika virus infection in three distinct cell types with two different

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service