Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2110

Sigma-Aldrich

WOBE437

≥98% (HPLC)

Synonym(s):

(2E,4E)-N-[2-(3,4-Dimethoxyphenyl)ethyl]dodeca-2,4-dienamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H33NO3
CAS Number:
Molecular Weight:
359.50
UNSPSC Code:
12352200
NACRES:
NA.77
Product SML2110 is currently not available for purchase. Contact Technical Service

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCCCCCC/C=C/C=C/C(NCCC1=CC(OC)=C(OC)C=C1)=O

InChI

1S/C22H33NO3/c1-4-5-6-7-8-9-10-11-12-13-22(24)23-17-16-19-14-15-20(25-2)21(18-19)26-3/h10-15,18H,4-9,16-17H2,1-3H3,(H,23,24)/b11-10+,13-12+

InChI key

LAHLFCSEHHJQRN-AQASXUMVSA-N

Biochem/physiol Actions

WOBE437 is a highly potent and selective endocannabinoid transport inhibitor. WOBE437 increases endocannabinoid levels in mice. It exhibits potent anxiolytic, anti-inflammatory, and analgesic effects in mice.
highly potent and selective endocannabinoid transport inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Chicca et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(25), E5006-E5015 (2017-06-07)
The extracellular effects of the endocannabinoids anandamide and 2-arachidonoyl glycerol are terminated by enzymatic hydrolysis after crossing cellular membranes by facilitated diffusion. The lack of potent and selective inhibitors for endocannabinoid transport has prevented the molecular characterization of this process

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service