Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML2030

Sigma-Aldrich

Tasimelteon

≥98% (HPLC)

Synonym(s):

(1R, 2R)-N-[2- (2,3-Dihydrobenzofuran-4-yl)cyclopropylmethyl]propanamide, BMS-214778, Hetlioz, MA 1, N-[[(1R,2R)-2-(2,3-Dihydro-4-benzofuranyl)cyclopropyl]methyl]propanamide, VEC-162

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19NO2
CAS Number:
Molecular Weight:
245.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(CC)NC[C@@H]1C[C@H]1C2=C3C(OCC3)=CC=C2

InChI

1S/C15H19NO2/c1-2-15(17)16-9-10-8-13(10)11-4-3-5-14-12(11)6-7-18-14/h3-5,10,13H,2,6-9H2,1H3,(H,16,17)/t10-,13+/m0/s1

InChI key

PTOIAAWZLUQTIO-GXFFZTMASA-N

Biochem/physiol Actions

Tasimelteon is a Dual Melatonin Receptor Agonist (DMRA) recently approved (Hetlioz) for the treatment of Non-24-hour sleep–wake disorder, a disorder primarily found in the totally blind, where a lack of normal synchronization between the 24-hour light–dark cycle and the endogenous circadian rhythm causes it to drift out of alignment with conventional sleep–wake schedules. The endogenous rhythm of the circadian pacemaker in the suprachiasmatic nuclei (SCN) is typically slightly longer than 24 h and therefore must be synchronized to the 24-hour day, normally by light, but exogenous melatonin can also phase-shift circadian rhythms. Tasimelteon is a selective agonist for the SCN melatonin receptors MT1 and MT2 with greater affinity for the MT2 receptor thought to be more important in mediating circadian rhythm phase-shifting. EC50 values are 0.75 nM for MT1 and 0.1 nM for MT2 measured by inhibition of forskolin-stimulated cAMP accumulation in NIH-3T3 cells expressing human MT1 and MT2.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wilbur P Trey Williams et al.
Pharmacotherapy, 36(9), 1028-1041 (2016-08-09)
Circadian rhythm sleep-wake disorders (CRSWDs) are characterized by persistent or recurrent patterns of sleep disturbance related primarily to alterations of the circadian rhythm system or the misalignment between the endogenous circadian rhythm and exogenous factors that affect the timing or
Christian Lavedan et al.
Neuropharmacology, 91, 142-147 (2014-12-24)
Hetlioz(®) (tasimelteon) is the first approved treatment in the United States for Non-24-Hour Sleep-Wake Disorder (Non-24). We present here data on the in vitro binding affinity of tasimelteon for both human melatonin receptors MT1 and MT2, as well as the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service