Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2028

Sigma-Aldrich

GA11

≥98% (HPLC)

Synonym(s):

2,6-Diphenylimidazo[1,2-a]pyridine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$139.00
25 MG
$450.00

$139.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$139.00
25 MG
$450.00

About This Item

Empirical Formula (Hill Notation):
C19H14N2
CAS Number:
Molecular Weight:
270.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$139.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

C12=NC(C3=CC=CC=C3)=CN1C=C(C4=CC=CC=C4)C=C2

InChI key

LETMYYZWCFHZLY-UHFFFAOYSA-N

Biochem/physiol Actions

GA11 is a BBB penetrant and potent inhibitor of ALDH
GA11 is a BBB penetrant and potent inhibitor of ALDH that inhibits the growth of mesenchymal (MES) glioma spheres. Also GA11 inhibits tumor growth in a murine GSC-derived xenograft model of glioblastoma.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service