Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1967

Sigma-Aldrich

N6-(4-Hydroxybenzyl) adenine riboside

≥98% (HPLC)

Synonym(s):

N-[(4-Hydroxyphenyl)methyl]adenosine, N6-(4-Hydroxybenzyl)-adenosine, NHBA, T1-11

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$129.00

$129.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$129.00

About This Item

Empirical Formula (Hill Notation):
C17H19N5O5
CAS Number:
Molecular Weight:
373.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$129.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](N2C=NC3=C2N=CN=C3NCC4=CC=C(O)C=C4)[C@H](O)[C@@H]1O

InChI

1S/C17H19N5O5/c23-6-11-13(25)14(26)17(27-11)22-8-21-12-15(19-7-20-16(12)22)18-5-9-1-3-10(24)4-2-9/h1-4,7-8,11,13-14,17,23-26H,5-6H2,(H,18,19,20)/t11-,13-,14-,17-/m1/s1

InChI key

UGVIXKXYLBAZND-LSCFUAHRSA-N

Biochem/physiol Actions

N6-(4-Hydroxybenzyl) adenine riboside (NHBA) is an orally available and brain penetrant active active ingredient of Gastrodia elata rhizomes used for the treatment of insomnia. N6-(4-Hydroxybenzyl) adenine riboside is an adenosine A2A receptor and the equilibrative nucleoside transporter 1 (ENT1) agonist. It exhibits sedative and hypnotic effects by binding to adenosine A1 and A2A receptors in mice. N6-(4-Hydroxybenzyl) adenine riboside exerted a therapeutic effect on HD transgenic mouse by decreasing protein level of polyglutamine-expanded huntingtin in the striatum.
Orally available and brain penetrant active adenosine A2A receptors and the equilibrative nucleoside transporter 1 (ENT1) agonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nai-Kuei Huang et al.
PloS one, 6(6), e20934-e20934 (2011-06-30)
Huntington's disease (HD) is a neurodegenerative disease caused by a CAG trinucleotide expansion in the Huntingtin (Htt) gene. The expanded CAG repeats are translated into polyglutamine (polyQ), causing aberrant functions as well as aggregate formation of mutant Htt. Effective treatments
Jhih-Bin Chen et al.
ChemMedChem, 6(8), 1390-1400 (2011-06-22)
A novel compound, N⁶-(4-hydroxybenzyl)adenosine, isolated from Gastrodia elata and which has been shown to be a potential therapeutic agent for preventing and treating neurodegenerative disease, was found to target both the adenosine A(2A) receptor (A(2A) R) and the equilibrative nucleoside
An-Hsun Chou et al.
Neuropharmacology, 99, 308-317 (2015-08-10)
More studies are required to develop therapeutic agents for treating spinocerebellar ataxia type 3 (SCA3), which is caused by mutant polyglutamine-expanded ataxin-3 and is the most prevalent subtype of spinocerebellar ataxias. T1-11 [N6-(4-Hydroxybenzyl) adenosine], isolated from a Chinese medicinal herb

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service