Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1937

Sigma-Aldrich

SN-6

≥98% (HPLC)

Synonym(s):

2-[4-(4-Nitrobenzyloxy)benzyl]thiazolidine-4-carboxylic acid ethyl ester, 4-Thiazolidinecarboxylic acid, 2-[[4-[(4-nitrophenyl)methoxy]phenyl]methyl]-, ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N2O5S
CAS Number:
Molecular Weight:
402.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OCC)C1CSC(CC2=CC=C(OCC3=CC=C([N+]([O-])=O)C=C3)C=C2)N1

InChI

1S/C20H22N2O5S/c1-2-26-20(23)18-13-28-19(21-18)11-14-5-9-17(10-6-14)27-12-15-3-7-16(8-4-15)22(24)25/h3-10,18-19,21H,2,11-13H2,1H3

InChI key

ZVYIJXLMBWCGHP-UHFFFAOYSA-N

Application

SN-6 has been used as a selective Na+/Ca++ exchanger (NCX) inhibitor to study its neuroprotective effects on axonal injury in mice.

Biochem/physiol Actions

SN-6 is a selective inhibitor of Na+/Ca++ exchanger (NCX) with some selectivity for the NCX1 isoform.
SN-6 is involved in safeguarding against hypoxia and reoxygenation‐induced cell damage in renal tubular cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satomi Kita et al.
Annals of the New York Academy of Sciences, 1099, 529-533 (2007-04-21)
We investigated the pharmacological properties of SN-6, a new selective Na+/Ca2+ exchanger (NCX) inhibitor. SN-6 preferentially inhibited the (45)Ca2+ uptake via NCX compared with the (45)Ca2+ efflux via NCX in NCX-transfected fibroblasts. SN-6 was three- to fivefold more inhibitory to
Anton Omelchenko et al.
Cell death & disease, 10(10), 727-727 (2019-09-29)
Mild traumatic brain injury (mTBI) is a frequently overlooked public health concern that is difficult to diagnose and treat. Diffuse axonal injury (DAI) is a common mTBI neuropathology in which axonal shearing and stretching induces breakdown of the cytoskeleton, impaired

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service