Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1862

Sigma-Aldrich

TP-472

≥98% (HPLC)

Synonym(s):

3-(6-Acetyl-pyrrolo[1,2-a]pyrimidin-8-yl)-N-cyclopropyl-4-methyl-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19N3O2
CAS Number:
Molecular Weight:
333.38
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C)C1=CC(C2=C(C)C=CC(C(NC3CC3)=O)=C2)=C4N=CC=CN41

Biochem/physiol Actions

TP-472 is a selective probe for bromodomain-containing proteins BRD9 and BRD7. This BRD9/7 probe was developed out of a collaborative effort by Takeda and the SGC, and is of a different chemotype from BRD9/7 probes previously available. TP-472 has a KD value of 33nM for BRD9 and 340nM for BRD7 by ITC. It is >30 fold selective for BRD9 over all other bromodomain family members except BRD7. It is suitable for in vivo applications. For full characterization details, please visit the TP-472 probe summary on the Structural Genomics Consortium (SGC) website.

TP-472N is used as a negative control and is available from Sigma. To learn more about and purchase TP-472N, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

TP-472 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service