Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1775

Sigma-Aldrich

K-756

≥98% (HPLC)

Synonym(s):

3-[[1-(6,7-Dimethoxy-4-quinazolinyl)-4-piperidinyl]methyl]-3,4-dihydro-2(1H)-quinazolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27N5O3
CAS Number:
Molecular Weight:
433.50
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C1NC2=C(C=CC=C2)CN1CC3CCN(C4=C(C=C(OC)C(OC)=C5)C5=NC=N4)CC3

Biochem/physiol Actions

K-756 is an orally available, potent and selective tankyrase inhibitor, which binds to the induced-pocket of tankyrase. K-756 is a selective Wnt/β-catenin pathway inhibitor that stabilizes Axin and reduces active β-catenin. K-756 inhibits the proliferation of APC mutant colorectal cancer COLO 320DM and SW403 cell lines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service