Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1746

Sigma-Aldrich

GS87

≥95% (HPLC)

Synonym(s):

4-[5-[[(3-Phenyl-1,2,4-oxadiazol-5-yl)methyl]thio]-1,3,4-oxadiazol-2-yl]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N5O2S
CAS Number:
Molecular Weight:
337.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C16H11N5O2S/c1-2-4-11(5-3-1)14-18-13(23-21-14)10-24-16-20-19-15(22-16)12-6-8-17-9-7-12/h1-9H,10H2

InChI key

HNTUWEZDVRVSFY-UHFFFAOYSA-N

Biochem/physiol Actions

GS87 is a highly specific GSK3 (glycogen synthase kinase 3) inhibitor that induces extensive differentiation of AML cells. GS87 potently inhibits growth of AML cells in mice with little effect on normal bone marrow cells. GS87 causes terminal differentiation of AML cells.
GS87 regulates target proteins, that participates in the multiplication and differentiation of cells, much efficiently than lithium and SB. It helps to initiate mitogen-activated protein kinase (MAPK) signaling, that is required for its differentiation activity. GS87 is considered as a candidate for clinical acute myeloid leukemia (AML) differentiation therapy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel glycogen synthase kinase-3 inhibitor optimized for acute myeloid leukemia differentiation activity
Hu S, et al.
Molecular Cancer Therapeutics, molcanther-molcan0566 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service