Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1721

Sigma-Aldrich

Vedaprofen

≥98% (HPLC)

Synonym(s):

(±)-2-(4-Cyclohexylnaphthalen-1-yl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22O2
CAS Number:
Molecular Weight:
282.38
Beilstein:
5568831
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(C(O)=O)c1ccc(C2CCCCC2)c3ccccc13

InChI

1S/C19H22O2/c1-13(19(20)21)15-11-12-16(14-7-3-2-4-8-14)18-10-6-5-9-17(15)18/h5-6,9-14H,2-4,7-8H2,1H3,(H,20,21)

InChI key

VZUGVMQFWFVFBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Vedaprofen is a non-steroidal anti-inflammatory drug (NSAID) that has been found to have antimicrobial activity. Vedaprofen is primarily used as a veterinary analgesic and anti-inflammatory for arthritis and pain. Its anti-inflammatory activity is due to cyclooxygenase inihbition, while its antimicrobial activity is less certain. Vedaprofen can kill B. subtilis by permeabilizing its membrane. Other studies have shown vedaprofen can target the Escherichia coli DNA polymerase III β subunit.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denise Tabacchi Fantoni et al.
BMC veterinary research, 11, 24-24 (2015-04-17)
This prospective randomized blinded clinical study aimed to investigate the potential of vedaprofen for preventive analgesia, comparing its analgesic effects with ketoprofen administered post-operatively in dogs undergoing maxillectomy or mandibulectomy. Pain control was effective and rescue analgesia was not necessary
Zhou Yin et al.
Chemistry & biology, 21(4), 481-487 (2014-03-19)
Evidence suggests that some nonsteroidal anti-inflammatory drugs (NSAIDs) possess antibacterial properties with an unknown mechanism. We describe the in vitro antibacterial properties of the NSAIDs carprofen, bromfenac, and vedaprofen, and show that these NSAIDs inhibit the Escherichia coli DNA polymerase III
Anne Lamsa et al.
ACS chemical biology, 11(8), 2222-2231 (2016-05-20)
Increasing antimicrobial resistance has become a major public health crisis. New antimicrobials with novel mechanisms of action (MOA) are desperately needed. We previously developed a method, bacterial cytological profiling (BCP), which utilizes fluorescence microscopy to rapidly identify the MOA of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service