Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1672

Sigma-Aldrich

JT010

≥98% (HPLC)

Synonym(s):

2-Chloro-N-(4-(4-methoxyphenyl)thiazol-2-yl)-N-(3-methoxypropyl)-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19ClN2O3S
CAS Number:
Molecular Weight:
354.85
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N(C1=NC(C2=CC=C(OC)C=C2)=CS1)CCCOC)CCl

InChI

1S/C16H19ClN2O3S/c1-21-9-3-8-19(15(20)10-17)16-18-14(11-23-16)12-4-6-13(22-2)7-5-12/h4-7,11H,3,8-10H2,1-2H3

InChI key

KZMAWJRXKGLWGS-UHFFFAOYSA-N

Biochem/physiol Actions

JT010 is a non-cytotoxic and highly stable (>90% remains after 2 h in 1 mM DTT or 2-mercaptoethanol) chloroacetamido warhead-carrying thiazole derivative that acts as a potent and TRPA1-selective channel activator via covalent modification of TRPA1 active site Cys621. JT0101 stimulates calcium influx in TRPA1-transfected HEK293 cells in a dose-dependent manner (EC50 = 650 pM) with excellent selectivity over TRPV1, TRPV3, TRPV4, TRPM2, TRPM8, and TRPC5 channels (EC50 >1 μM).
JT010 is a non-cytotoxic and highly stable chloroacetamido warhead-carrying thiazole derivative.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kento Tazawa et al.
The American journal of pathology, 190(12), 2417-2426 (2020-09-14)
Increased expression of the transient receptor potential ankyrin 1 (TRPA1) channel has been detected in carious tooth pulp, suggesting involvement of TRPA1 in defense or repair of this tissue after exogenous noxious stimuli. This study aimed to elucidate the induction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service