Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1628

Sigma-Aldrich

LMT-28

≥98% (HPLC)

Synonym(s):

(4S)-3-[(2S,3S)-3-Hydroxy-2-methyl-4-methylene-1-oxononyl]-4-(1-methylethyl)-2-oxazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H29NO4
CAS Number:
Molecular Weight:
311.42
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

oil

color

colorless to light yellow, oil

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(C)[C@H](N1C([C@@H](C)[C@H](O)C(CCCCC)=C)=O)COC1=O

InChI

1S/C17H29NO4/c1-6-7-8-9-12(4)15(19)13(5)16(20)18-14(11(2)3)10-22-17(18)21/h11,13-15,19H,4,6-10H2,1-3,5H3/t13-,14+,15+/m0/s1

InChI key

UDXWSYOXIRPYFK-RRFJBIMHSA-N

Biochem/physiol Actions

LMT-28 is a derivative of oxazolidinone. It has the ability to repress the activation of signal transducer and activator of transcription 3 (STAT3) stimulated by interleukin 6 (IL-6). Orally administered LMT-28 reduced arthritis and acute pancreatitis stimulated by collagen in mice pathologic models.
LMT-28 is an IL-6 antagonist that targets the IL-6 Receptor β subunit, Glycoprotein 130 (gp130).
LMT-28 is an IL-6 antagonist that targets the IL-6 Receptor β subunit, Glycoprotein 130 (gp130). Its binding to gp130) results in inhibition of IL-6 activation, and downregulation of levels of p-STAT3. LMT-28 was shown to stimulate phosphorylation of STAT3 and JAK2 protein. IL-6 dysregulation is involved in inflammation and cancers. LMT-28 had inhbitory activity against IL-6–dependent TF-1 cell proliferation and also had therapeutic effect against pancreatitis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A Novel Small-Molecule Inhibitor Targeting the IL-6 Receptor β Subunit, Glycoprotein 130.
Hong SS, et al.
Journal of Immunology, 195(1), 45-237 (2015)
Mónica Romera-Hernández et al.
Cell reports, 30(1), 37-45 (2020-01-09)
Tissue repair requires temporal control of progenitor cell proliferation and differentiation to replenish damaged cells. In response to acute insult, group 3 innate lymphoid cells (ILC3s) regulate intestinal stem cell maintenance and subsequent tissue repair. ILC3-derived IL-22 is important for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service