Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1607

Sigma-Aldrich

Lesinurad

≥98% (HPLC)

Synonym(s):

2-[[5-Bromo-4-(4-cyclopropyl-1-naphthalenyl)-4H-1,2,4-triazol-3-yl]thio]-acetic acid, RDEA 594

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$157.00
50 MG
$627.00

$157.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$157.00
50 MG
$627.00

About This Item

Empirical Formula (Hill Notation):
C17H14BrN3O2S
CAS Number:
Molecular Weight:
404.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$157.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

−20°C

SMILES string

BrC1=NN=C(SCC(O)=O)N1C2=C3C(C=CC=C3)=C(C4CC4)C=C2

InChI

1S/C17H14BrN3O2S/c18-16-19-20-17(24-9-15(22)23)21(16)14-8-7-11(10-5-6-10)12-3-1-2-4-13(12)14/h1-4,7-8,10H,5-6,9H2,(H,22,23)

InChI key

FGQFOYHRJSUHMR-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Lesinurad is a selective inhibitor of Urate transporter 1 (URAT1), a urate-anion exchanger which is responsible for the majority of uric acid reabsorption from the renal tubular lumen. Lesinurad thus reduces serum uric acid levels. Lesinurad has been approved for use in combination with a xanthine oxidase inhibitor to treat hyperuricemia associated with gout.
Lesinurad is a selective inhibitor of Urate transporter 1 (URAT1).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service