Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1487

Sigma-Aldrich

ABT-089 dihydrochloride

≥98% (HPLC)

Synonym(s):

2-methyl-3-(2-(S)-pyrrolidinylmethoxy)pyridine dihydrochloride, Pozanicline dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O · 2HCl
CAS Number:
Molecular Weight:
265.18
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +20 to +30°, c = 1 in methanol

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(OC[C@@H]2CCCN2)C=CC=N1.[H]Cl.[H]Cl

InChI

1S/C11H16N2O.2ClH/c1-9-11(5-3-6-12-9)14-8-10-4-2-7-13-10;;/h3,5-6,10,13H,2,4,7-8H2,1H3;2*1H/t10-;;/m0../s1

InChI key

UGVSESCRKJTJAK-XRIOVQLTSA-N

Biochem/physiol Actions

ABT-089 (Pozanicline) is a subtype-selective neuronal nicotinic receptor partial agonist binding primarily to the α4β2 and also α6β2 subtypes. It has been investigated for alleviating symptoms of nicotine withdrawal including cognitive deficits and anxiety.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service