Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1439

Sigma-Aldrich

ML354

≥98% (HPLC)

Synonym(s):

1-Methyl-5-nitro-3-phenyl-1H-indole-2-methanol, VU0099704

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2O3
CAS Number:
Molecular Weight:
282.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

H2O: 10 mg/mL, clear
DMSO: 25 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

CN(C(CO)=C1C2=CC=CC=C2)C3=C1C=C([N+]([O-])=O)C=C3

InChI

1S/C16H14N2O3/c1-17-14-8-7-12(18(20)21)9-13(14)16(15(17)10-19)11-5-3-2-4-6-11/h2-9,19H,10H2,1H3

InChI key

GNJUKVGDCUKDLF-UHFFFAOYSA-N

Biochem/physiol Actions

ML354 is a potent and selective PAR4 (protease activated receptor-4) antagonist.

Other Notes

Light and air sensitive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service