Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1419

Sigma-Aldrich

B32B3

≥95% (HPLC)

Synonym(s):

1H-Indole-3-carboxaldehyde 2-(5,6,7,?8-tetrahydro[1]benzothieno[2,3-d]pyrimidin-4-yl)hydrazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N5S
CAS Number:
Molecular Weight:
347.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

light yellow to dark yellow

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

C1(C(NN=CC2=CNC3=C2C=CC=C3)=NC=N4)=C4SC5=C1CCCC5

InChI

1S/C19H17N5S/c1-3-7-15-13(5-1)12(9-20-15)10-23-24-18-17-14-6-2-4-8-16(14)25-19(17)22-11-21-18/h1,3,5,7,9-11,20H,2,4,6,8H2,(H,21,22,24)

InChI key

AKDKHZVFMAWBFX-UHFFFAOYSA-N

Biochem/physiol Actions

B32B3 is a potent, selective and cell permeable VprBP inhibitor that suppresses xenograft tumor growth. B32B3 potently inhibits VprBP dependent formation of phosphorylated histone H2A on threonine 120 (H2AT120p).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service