Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1376

Sigma-Aldrich

BTC

≥95% (HPLC)

Synonym(s):

2-(4-Hydroxyphenyl)-1-benzothiophene-6-ol, 2-(4-Hydroxyphenyl)benzo[b]thiophene-6-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O2S
CAS Number:
Molecular Weight:
242.29
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

storage condition

protect from light

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

InChI

1S/C14H10O2S/c15-11-4-1-9(2-5-11)13-7-10-3-6-12(16)8-14(10)17-13/h1-8,15-16H

InChI key

MDGWZLQPNOETLH-UHFFFAOYSA-N

Biochem/physiol Actions

BTC is a selective estrogen receptor (ER) modulator (SERM) that acts as a selective estrogen mimic (SEM). BTC has been found to be both estrogenic and antiproliferative in ER-positive endocrine-resistant breast cancer cell lines that overexpress protein kinase C alpha (PKCα), which is often associated with tamoxifen resistance. BTC caused regression of endocrine-resistant breast cancer xenografts, and did not support the growth of hormone-dependent xenografts in vivo or cause an increase in uterine weight. As in the case with raloxifene and estradiol, BTC tumor regression was accompanied by the exit of ERα from the nucleus, suggesting extranuclear ERα may be involved in the mehanism of action.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fuping Li et al.
Small (Weinheim an der Bergstrasse, Germany), 16(14), e1906276-e1906276 (2020-03-05)
Direct conversion of syngas to dimethyl ether (DME) through the intermediate of methanol allows more efficient DME production in a simpler reactor design relative to the conventional indirect route. Although Cu/ZnO-based multicomponent catalysts are highly active for methanol synthesis in
Rui Pei et al.
Journal of hazardous materials, 384, 121418-121418 (2019-12-11)
Three-dimensional (3D) printing technique has received exceptional global attention as it can create a myriad of high-resolution architectures from digital models. In the present study, 3D-printed metal-organic frameworks (MOFs) were shaped into several geometries via direct ink writing, which overcomes
Xiaozhou Huang et al.
Analytical and bioanalytical chemistry, 412(4), 849-860 (2020-01-04)
In this paper, a novel bimetallic Ce-Ni metal-organic frameworks (Ce-Ni-MOF) are synthesized by hydrothermal reaction, using 1,3,5-benzenetricarboxylic acid as a ligand. In particular, the bimetallic Ce-Ni-MOF with the largest specific surface area and catalytic sites was synthesized when the molar
Xinyang Zhang et al.
The Analyst, 145(14), 5011-5018 (2020-06-11)
3D magnetic porous carbon (MPCK) was prepared using the metal-organic framework (MOF) MIL-100(Fe) as the carbon precursor by carbonisation and KOH activation strategies. Carbonisation and activation ensured that MPCK possessed excellent structural and thermal stability, strong magnetic responsiveness and high
Axel X Canatelli et al.
ChemPlusChem, 85(10), 2349-2356 (2020-10-24)
Metal-organic frameworks (MOFs) are known for their versatility in terms of their crystalline structure, porosity, resistance to temperature, radiation damage, and luminescence among others. Gadolinium (Gd) is one of the elements with the highest reported cross-section for low energy neutron

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service