Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1340

Sigma-Aldrich

ML193 trifluoroacetate

≥98% (HPLC)

Synonym(s):

AC1LOE9L, CID-1261822, CID1261822, N-{4-[(3,4-Dimethyl-1,2-oxazol-5-yl)sulfamoyl]phenyl}-6,8-dimethyl-2-(pyridin-2-yl)quinoline-4-carboxamide trifluoroacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$136.00
25 MG
$657.00

$136.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$136.00
25 MG
$657.00

About This Item

Empirical Formula (Hill Notation):
C28H25N5O4S · C2HF3O2
CAS Number:
Molecular Weight:
641.62
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$136.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(C(O)=O)F.CC1=C(ON=C1C)NS(C2=CC=C(NC(C3=CC(C4=CC=CC=N4)=NC5=C3C=C(C)C=C5C)=O)C=C2)(=O)=O

InChI

1S/C28H25N5O4S.C2HF3O2/c1-16-13-17(2)26-22(14-16)23(15-25(31-26)24-7-5-6-12-29-24)27(34)30-20-8-10-21(11-9-20)38(35,36)33-28-18(3)19(4)32-37-28;3-2(4,5)1(6)7/h5-15,33H,1-4H3,(H,30,34);(H,6,7)

InChI key

VLRPKRWNDZQTPS-UHFFFAOYSA-N

Biochem/physiol Actions

ML193 is a potent and selective piperadinyloxadiazolone antagonist for G protein-coupled receptor (GPCR) GPR55, a receptor for L-α-lysophosphatidylinositol (LPI) which also binds synthetic cannabinoids, and signals through several endogenous pathways. GPR55 is highly abundant in the CNS as well as in intestine, bone marrow, spleen, platelets, immune and endothelial cells. Its role is not completely understood, but has been implicated in inflammatory pain, neuropathic pain, metabolic disorder, bone development, and cancer. Selective antagonists are needed to further elucidate the role of GPR55 in physiology and pathobiology. ML193 (CID1261822) has 221 nM potency for GPR55 and >145-fold, >27-fold and >145-fold antagonist selectivity against GPR35, cannabinoid receptors CB1 and CB2, respectively and >145-fold agonist selectivity against GPR35, CB1 and CB2. ML193 is inhibits the downstream responses of ERK phosphorylation with an IC50 of 65 nM and PKC βII translocation with an IC50 of 10 μM.
ML193 is a potent and selective piperadinyloxadiazolone antagonist for G protein-coupled receptor (GPCR) GPR55.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justine Yu et al.
The Journal of biological chemistry, 288(31), 22481-22492 (2013-07-03)
The L-α-lysophosphatidylinositol (LPI)-sensitive receptor GPR55 is coupled to Ca(2+) signaling. Low levels of GPR55 expression in the heart have been reported. Similar to other G protein-coupled receptors involved in cardiac function, GPR55 may be expressed both at the sarcolemma and
Evangelia Kotsikorou et al.
Biochemistry, 52(52), 9456-9469 (2013-11-28)
GPR55 is a class A G protein-coupled receptor (GPCR) that has been implicated in inflammatory pain, neuropathic pain, metabolic disorder, bone development, and cancer. Initially deorphanized as a cannabinoid receptor, GPR55 has been shown to be activated by non-cannabinoid ligands
Sandra Cecconi et al.
International journal of molecular sciences, 20(12) (2019-06-20)
Endocannabinoids are key-players of female fertility and potential biomarkers of reproductive dysfunctions. Here, we investigated localization and expression of cannabinoid receptor type-1 and -2 (CB1R and CB2R), G-protein coupled receptor 55 (GPR55), and transient receptor potential vanilloid type 1 channel

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service