Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1296

Sigma-Aldrich

6-Thio-2′-Deoxyguanosine

≥97% (HPLC)

Synonym(s):

2′-Ddeoxy-6-thio-guanosine, 2′-Deoxy-6-thioguanosine, 2′-Deoxythioguanosine, 2-Amino-9-(2-deoxy-β-D-erythro-pentofuranosyl)-6-mercaptopurine, 2-Amino-9-(2-deoxy-β-D-erythro-pentofuranosyl)-9H-purine-6(1H)-thione (7CI,8CI), 6-Thio-dG, NSC 71261

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O3S
CAS Number:
Molecular Weight:
283.31
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

S=C1C2=C(N([C@H]3C[C@H](O)[C@@H](CO)O3)C=N2)N=C(N)N1

InChI

1S/C10H13N5O3S/c11-10-13-8-7(9(19)14-10)12-3-15(8)6-1-4(17)5(2-16)18-6/h3-6,16-17H,1-2H2,(H3,11,13,14,19)/t4-,5-,6-/m1/s1

InChI key

SCVJRXQHFJXZFZ-HSUXUTPPSA-N

Application

6-Thio-2′-Deoxyguanosine has been used in secreted luciferase enzyme activity in tonsillar cancer cells.

Biochem/physiol Actions

6-Thio-2′-deoxyguanosine (6-thio-dG) is a nucleoside analog and telomerase substrate that is incorporated into de novo–synthesized telomeres. 6-Thio-2′-deoxyguanosine induces telomere dysfunction and rapid cell death in cancer cells, while spearing telomerase-negative cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haoran Yu et al.
Virology, 521, 1-19 (2018-06-05)
Adenosine plays an important role in cell death and differentiation as well as in tumorigenesis and the intra- and extra-cellular levels range from nanomolar to millimolar levels under various physiological or pathophysiological conditions. Here we report that adenosine can activate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service