Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1258

Sigma-Aldrich

ML099

≥98% (HPLC)

Synonym(s):

2-[(2-Phenylethyl)thio]-3-pyridinecarboxylic acid, CID-888706

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13NO2S
CAS Number:
Molecular Weight:
259.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C1=CC=CN=C1SCC(C2=CC=CC=C2)=O)=O

InChI

1S/C14H11NO3S/c16-12(10-5-2-1-3-6-10)9-19-13-11(14(17)18)7-4-8-15-13/h1-8H,9H2,(H,17,18)

InChI key

MJHMABNIGSEYKN-UHFFFAOYSA-N

Biochem/physiol Actions

ML099 is a Ras-related GTPase activator. Mutated forms of small GTPases induce proliferation and transformation of a number of cell types as well as differentiation of neuronal cells. The only known activation mechanism of small GTPase is via guanine nucleotide-exchange factors (GEFs). Data suggests that some diseases, including immunodeficienty syndromes result from loss of function mutations in the Rac2 GTPase, and the GEF cdc42. ML099 can activate cdc42, Rac1 and Ras GTPase activity with mid to high nM EC50s. GTPase activatiors could be useful tools for defining the mechanism of GEF activation of GTPases, further understanding of GTPase signaling mechanisms and study of diseases in which loss of function mutants are implicated.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service