Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1161

Sigma-Aldrich

Ki11502

≥98% (HPLC)

Synonym(s):

Ki 11502, N-[[[4-[(6,7-Dimethoxy-4-quinolinyl)oxy]phenyl]amino]thioxomethyl]-2-methyl-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H23N3O4S
CAS Number:
Molecular Weight:
473.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 20 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

S=C(NC(C1=C(C)C=CC=C1)=O)NC(C=C2)=CC=C2OC3=CC=NC4=CC(OC)=C(OC)C=C43

InChI

1S/C26H23N3O4S/c1-16-6-4-5-7-19(16)25(30)29-26(34)28-17-8-10-18(11-9-17)33-22-12-13-27-21-15-24(32-3)23(31-2)14-20(21)22/h4-15H,1-3H3,(H2,28,29,30,34)

InChI key

ZXGIBSBJQLLUEE-UHFFFAOYSA-N

Biochem/physiol Actions

Ki11502 is a potent, ATP-competitive multikinase inhibitor that is selective for PDGF receptor. Ki11502 inhibits malignant hematopoietic cells proliferation and induces their apoptosis. Ki11502 is effective against colorectal cancer SW480 cell line.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service