Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1131

Sigma-Aldrich

Fosbretabulin disodium

≥98% (HPLC)

Synonym(s):

2-Methoxy-5-[(1Z)-2-(3,4,5-trimethoxyphenyl)ethenyl]phenyl disodium phosphate, CA4P, Combretastatin A4 phosphate disodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19O8P · 2Na
CAS Number:
Molecular Weight:
440.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 5 mg/mL, clear

storage temp.

−20°C

SMILES string

[O-]P(OC1=CC(/C=C\C2=CC(OC)=C(OC)C(OC)=C2)=CC=C1OC)([O-])=O.[Na+].[Na+]

InChI

1S/C18H21O8P.2Na/c1-22-14-8-7-12(9-15(14)26-27(19,20)21)5-6-13-10-16(23-2)18(25-4)17(11-13)24-3;;/h5-11H,1-4H3,(H2,19,20,21);;/q;2*+1/p-2/b6-5-;;

InChI key

VXNQMUVMEIGUJW-XNOMRPDFSA-L

Biochem/physiol Actions

Fosbretabulin disodium (Combretastatin A4 phosphate, CA4P) is a potent vascular-disrupting agent (VDA). Fosbretabulin disodium is a prodrug that is converted to combretastatin A (Sigma Cat No. C7744) inside the endothelial cells that line blood vessels, where it binds to tubulin dimers and prevents microtubule polymerization, resulting in mitotic arrest and apoptosis in endothelial cells. Fosbretabulin disodium is a potent anti-cancer agent that exhibits antivascular effects on tumor vasculature, inducing a rapid reduction in tumor blood flow and a concomitant increase of cellular necrosis. Some of its activity is believed to involve interference with vascular endothelial-cadherin signaling in addition to its microtubule-disrupting activity.
Fosbretabulin disodium is a vascular disrupting agent; prodrug that is converted to combretastatin A inside the endothelial cells.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Fan et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 121, 74-84 (2018-05-18)
YMR-65​, 5-(5-bromo-1-methyl-1H-indol-3-yl)-3-(3-methoxyphenyl)-4, 5-dihydro-1H-pyrazole-1-carboxamide, is a potential tubulin inhibitor exhibiting good anticancer activity. In our study, we illustrated the biological activities in HepG2 cells and the pharmacodynamic and pharmacokinetic profiles were evaluated in murine H22 hepatoma-bearing mice. Molecular docking assay and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service