Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0831

Sigma-Aldrich

ML261

≥98% (HPLC)

Synonym(s):

2-Chloro-N-[2-(3,4-dimethoxyphenyl)ethyl]-4-methyl-4H-thieno[3,2-b]pyrrole-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23ClN2O3S
CAS Number:
Molecular Weight:
406.93
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

ML261 blocks lipid droplet formation in murine AML-12 hepatocytes grown in the presence of oleic acid, with an IC50 of 69.7 nM. ML261 appears to be species specific, the compound does not affect lipid droplet formation in HuH7 or primary human hepatocytes.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service