Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0696

Sigma-Aldrich

transpDMA

≥98% (HPLC)

Synonym(s):

4-[(4R,6R)-4,6-Diphenyl-1,3-dioxan-2-yl]-N,N-dimethyl-benzenamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H25NO2
CAS Number:
Molecular Weight:
359.46
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

transpDMA is a constitutive androstane receptor (CAR) agonist that inhibits FOXO1 and HNF4 driven expression of the gluconeogeneis PEPCK and glucose-6- phosphatase. transpDMA reduces blood glucose levels, insulin sensitivity and weight gain in rats fed a high fat diet.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service