Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0665

Sigma-Aldrich

LY-456236

≥98% (HPLC)

Synonym(s):

(4-Methoxy-phenyl)-(6-methoxy-quinazolin-4-yl)-amine hydrochloride, 6-Methoxy-N-(4-methoxyphenyl)-4-quinazolinamine hydrochloride, LY456236, MPMQ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N3O2 · HCl
CAS Number:
Molecular Weight:
317.77
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C16H15N3O2.ClH/c1-20-12-5-3-11(4-6-12)19-16-14-9-13(21-2)7-8-15(14)17-10-18-16;/h3-10H,1-2H3,(H,17,18,19);1H

InChI key

AVKFOWUSTVWZQN-UHFFFAOYSA-N

Biochem/physiol Actions

LY-456236 is a selective mGlu1 receptor antagonist with an IC50 value of 143 nM for mGlu1 compared to > 10 μM for mGlu5 receptors.
LY-456236 is a selective mGlu1 receptor antagonist.

Features and Benefits

This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service