Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0634

Sigma-Aldrich

CID2858522

≥98% (HPLC)

Synonym(s):

1-[3,5-Bis(1,1-dimethylethyl)-4-hydroxyphenyl]-2-[2-[(3-hydroxypropyl)amino]-5,6-dimethyl-1H-benzimidazol-1-yl]-ethanone

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$386.00

$386.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$386.00

About This Item

Empirical Formula (Hill Notation):
C28H39N3O3
CAS Number:
Molecular Weight:
465.63
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$386.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Cc1cc2nc(NCCCO)n(CC(=O)c3cc(c(O)c(c3)C(C)(C)C)C(C)(C)C)c2cc1C

InChI

1S/C28H39N3O3/c1-17-12-22-23(13-18(17)2)31(26(30-22)29-10-9-11-32)16-24(33)19-14-20(27(3,4)5)25(34)21(15-19)28(6,7)8/h12-15,32,34H,9-11,16H2,1-8H3,(H,29,30)

InChI key

CYCGGKILBWERDJ-UHFFFAOYSA-N

Biochem/physiol Actions

CID2858522 specifically inhibits NF-kB activation downstream of protein kinsase C (PKC).
CID2858522 specifically inhibits NF-kB activation downstream of protein kinsase C (PKC). CID2858522 dose dependently inhibits NF-kB reporter activity in PMA-ionomycin stimulated HEK293 cells, but does not affect NF-kB activity in cells challenged with TNFa, retinoic acid, or stimulation of the toll-like receptor signaling pathway.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandria P Harrold et al.
European journal of pharmacology, 873, 172981-172981 (2020-02-06)
Dysregulated activity of the transcription factors of the nuclear factor κb (NF-κB) family has been implicated in numerous cancer types, inflammatory diseases, autoimmune disease, and other disorders. As such, selective NF-κB pathway inhibition is an attractive target to researchers for

Articles

Protein kinase C (PKC) is an AGC kinase that phosphorylates serine and threonine residues in many target proteins.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service