Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0615

Sigma-Aldrich

Br-DIF-1

≥98% (HPLC)

Synonym(s):

1-(3,5-Dibromo-2,6-dihydroxy-4-methoxyphenyl)-1-Hexanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16Br2O4
CAS Number:
Molecular Weight:
396.07
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Br-DIF-1 is a bromine-substituted derivative of DIF-1 (differentiation-inducing factor-1).
Br-DIF-1 is a bromine-substituted derivative of DIF-1 (differentiation-inducing factor-1). Br-DIF-1 significantly increases the efficiency of DMSO-induced differentiation of P19CL6 cells into beating cardiomyocytes by maintaing expression levels of the T-type calcium channel Cav3.1.

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to calcium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service