Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0595

Sigma-Aldrich

A-286982

≥95% (HPLC)

Synonym(s):

(2E)-1-(4-Acetyl-1-piperazinyl)-3-[4-[[2-(1-methylethyl)phenyl]thio]-3-nitrophenyl]-2-propen-1-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$122.00

$122.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$122.00

About This Item

Empirical Formula (Hill Notation):
C24H27N3O4S
CAS Number:
Molecular Weight:
453.55
UNSPSC Code:
12352200
NACRES:
NA.77

$122.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

S(c2c(cc(cc2)\C=C\C(=O)N3CCN(CC3)C(=O)C)[N+](=O)[O-])c1c(cccc1)C(C)C

InChI

1S/C24H27N3O4S/c1-17(2)20-6-4-5-7-22(20)32-23-10-8-19(16-21(23)27(30)31)9-11-24(29)26-14-12-25(13-15-26)18(3)28/h4-11,16-17H,12-15H2,1-3H3/b11-9+

InChI key

HTGGAYLWTDOFDK-PKNBQFBNSA-N

Biochem/physiol Actions

A-286982 blocks binding of lymphocyte function-associated antigen-1 (LFA-1) with the cell adhesion molecule ICAM-1.
A-286982 blocks binding of lymphocyte function-associated antigen-1 (LFA-1) with the cell adhesion molecule ICAM-1. The IC50 in a cell based assay is 35 nM.

Other Notes

Air sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service