Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0566

Sigma-Aldrich

VU0359595

≥97% (HPLC)

Synonym(s):

(1R,2R)-N-([S]-1-{4-[5-Bromo-2-oxo-2,3-dihydro-1H-benzo(d)imidazol-1-yl]piperidin-1-yl}propan-2-yl)-2-phenylcyclopropanecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H29BrN4O2
CAS Number:
Molecular Weight:
497.43
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N(C2CCN(C[C@H](C)NC([C@H]3[C@H](C4=CC=CC=C4)C3)=O)CC2)C5=CC=C(Br)C=C5N1

InChI

1S/C25H29BrN4O2/c1-16(27-24(31)21-14-20(21)17-5-3-2-4-6-17)15-29-11-9-19(10-12-29)30-23-8-7-18(26)13-22(23)28-25(30)32/h2-8,13,16,19-21H,9-12,14-15H2,1H3,(H,27,31)(H,28,32)/t16-,20-,21+/m0/s1

InChI key

JSVNNLRZCJAYTQ-ORYQWCPZSA-N

Biochem/physiol Actions

VU0359595 is a PLD1 specific inhibitor with an IC50 of 3.7 nM, and 1500-fold selectivity over PLD2 (IC50 = 6.4 μM).
VU0359595 is a PLD1 specific inhibitor.

Features and Benefits

This compound is featured on the Phospholipase D page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuelin Han et al.
PloS one, 6(7), e21468-e21468 (2011-07-16)
The internalization of Aspergillus fumigatus into lung epithelial cells is a process that depends on host cell actin dynamics. The host membrane phosphatidylcholine cleavage driven by phospholipase D (PLD) is closely related to cellular actin dynamics. However, little is known

Articles

PLD enzymes catalyze PC hydrolysis, influencing substrate recognition and lipid metabolism.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service