Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0510

Sigma-Aldrich

S 26948

≥98% (HPLC)

Synonym(s):

2-[[4-[2-(6-Benzoyl-2-oxo-3(2H)-benzothiazolyl)ethoxy]phenyl]methyl]-propanedioic acid 1,3-dimethyl ester, S26948

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$137.00
25 MG
$505.00

$137.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$137.00
25 MG
$505.00

About This Item

Empirical Formula (Hill Notation):
C28H25NO7S
CAS Number:
Molecular Weight:
519.57
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$137.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

COC(=O)C(Cc1ccc(OCCN2C(=O)Sc3cc(ccc23)C(=O)c4ccccc4)cc1)C(=O)OC

InChI

1S/C28H25NO7S/c1-34-26(31)22(27(32)35-2)16-18-8-11-21(12-9-18)36-15-14-29-23-13-10-20(17-24(23)37-28(29)33)25(30)19-6-4-3-5-7-19/h3-13,17,22H,14-16H2,1-2H3

InChI key

NSMJEHGOMXSLCW-UHFFFAOYSA-N

Biochem/physiol Actions

S 26948 belongs to the non-thiazolidinedione (TZD) class of drugs. It has the potential to treat diabetes and atherogenesis.[1]
S 26948 is a partial PPARγ agonist; selective PPARγ modulator (SPPARγM).
S 26948 is a selective PPARγ modulator (SPPARγM). S 26948 is as effective as rosiglitazone in reducing insulin resistance and lipid homeostasis, but does not increase body or white adipose tissue weight. S 26948 also leads to different co-activatior recruitment to PPARγ than rosiglitazone.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) and Nuclear Receptors (PPARs) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathryn A Crawford et al.
Aquatic toxicology (Amsterdam, Netherlands), 210, 30-43 (2019-03-02)
Sentinel species such as the Atlantic killifish (Fundulus heteroclitus) living in urban waterways can be used as toxicological models to understand impacts of environmental metabolism disrupting compound (MDC) exposure on both wildlife and humans. Exposure to MDCs is associated with
Maria Carmen Carmona et al.
Diabetes, 56(11), 2797-2808 (2007-08-21)
Rosiglitazone displays powerful antidiabetes benefits but is associated with increased body weight and adipogenesis. Keeping in mind the concept of selective peroxisome proliferator-activated receptor (PPAR)gamma modulator, the aim of this study was to characterize the properties of a new PPARgamma

Articles

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

We offer many products related to non-steroid nuclear receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service