Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0500

Sigma-Aldrich

GSK334429 hydrochloride

≥98% (HPLC)

Synonym(s):

1-(1-Methylethyl)-4-((1-(6-(trifluoromethyl)-3-pyridinyl)-4-piperidinyl)carbonyl)hexahydro-1H-1,4-diazepine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$195.00

$195.00


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
$195.00

About This Item

Empirical Formula (Hill Notation):
C20H29F3N4O · xHCl
CAS Number:
Molecular Weight:
398.47 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$195.00


Please contact Customer Service for Availability

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 5 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

Cl.CC(C)N1CCCN(CC1)C(=O)C2CCN(CC2)c3ccc(nc3)C(F)(F)F

InChI

1S/C20H29F3N4O.ClH/c1-15(2)25-8-3-9-27(13-12-25)19(28)16-6-10-26(11-7-16)17-4-5-18(24-14-17)20(21,22)23;/h4-5,14-16H,3,6-13H2,1-2H3;1H

InChI key

RCZVWLOXPFGZDD-UHFFFAOYSA-N

Biochem/physiol Actions

GSK334429 is a selective histamine H3 receptor antagonist.
GSK334429 is a selective histamine H3 receptor antagonist. H3 antagonists increase the release of brain histamine, ACh, NE and DA neurotransmitters that modulate cognitive processes. Studies look at their analgesic efffects including efficacy for neuropathic pain.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Store desiccated; dry room.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to histamine receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service