Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML0489

Sigma-Aldrich

BI-87G3

≥98% (HPLC)

Synonym(s):

2-(5-Nitrothiazol-2-ylthio)benzo[d]thiazole, 2-[(5-Nitro-2-thiazolyl)thio]-benzothiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5N3O2S3
CAS Number:
Molecular Weight:
295.36
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1cnc(Sc2nc3ccccc3s2)s1

InChI

1S/C10H5N3O2S3/c14-13(15)8-5-11-9(17-8)18-10-12-6-3-1-2-4-7(6)16-10/h1-5H

InChI key

UHBFMJAKLZGVDS-UHFFFAOYSA-N

Biochem/physiol Actions

BI-87G3 is a cell-permeable, potent and selective competitive inhibitor of the c-Jun N-terminal kinase (JNK). BI-87G3 is a JNK inhibitor that targets the JIP-JNK interaction site.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Garthiga Manickam et al.
Molecular and cellular biology, 39(4) (2018-12-12)
Sphingomyelin phosphodiesterase 3 (SMPD3), a lipid-metabolizing enzyme present in bone and cartilage, has important roles in the developing skeleton. We previously showed that SMPD3 deficiency results in delayed extracellular matrix (ECM) mineralization and severe skeletal deformities in an inducible knockout
Corina A Schmid et al.
The Journal of experimental medicine, 212(5), 775-792 (2015-04-08)
The epigenetic dysregulation of tumor suppressor genes is an important driver of human carcinogenesis. We have combined genome-wide DNA methylation analyses and gene expression profiling after pharmacological DNA demethylation with functional screening to identify novel tumor suppressors in diffuse large

Articles

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service