Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0455

Sigma-Aldrich

1,2,3,4,5,6-Hexabromocyclohexane

≥98% (HPLC)

Synonym(s):

1,2,3,4,5,6-Hexabromocyclohexane, Benzene hexabromide, NSC 7908

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Br6
CAS Number:
Molecular Weight:
557.54
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL (clear solution, warmed)

storage temp.

room temp

SMILES string

BrC1C(Br)C(Br)C(Br)C(Br)C1Br

InChI

1S/C6H6Br6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H

InChI key

QFQZKISCBJKVHI-UHFFFAOYSA-N

Biochem/physiol Actions

1,2,3,4,5,6-hexabromocyclohexane binds directly to a pocket within the kinase domain of JAK2 and inhibits autophosphorylation. The compound also blocks growth hormone induced JAK2 autophosphorylation without affecting EFGR autophosphorylation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kotaro Horiguchi et al.
Cell and tissue research, 364(2), 395-404 (2015-11-18)
Secretion of hormones by the anterior pituitary gland can be stimulated or inhibited by paracrine factors that are produced during inflammatory reactions. The inflammation cytokine interferon-gamma (IFN-γ) is known to inhibit corticotropin-releasing factor (CRF)-stimulated adrenocorticotropin (ACTH) release but its signaling
Jeffrey D Kittilson et al.
Frontiers in endocrinology, 2, 30-30 (2011-01-01)
Previously, we found that the teleost fish, rainbow trout, possesses two growth hormone receptor (GHR) subtypes that display distinct ligand-binding and agonist-induced regulation features. In this study, we used Chinese hamster ovary-K1 cells stably transfected individually with the two trout
Anshu Agrawal et al.
Aging, 4(9), 648-659 (2012-10-16)
Advancing age leads to significant decline in immune functions. IL-21 is produced primarily by T follicular helper (Tfh) cells and is required for effective immune cell functions. Here we compared the induction of IL-21 in aged and young subjects. Our
Xue Jiang et al.
Frontiers in endocrinology, 11, 20-20 (2020-02-23)
In mammals, local production of tumor necrosis factor α (TNFα) inhibits growth hormone (GH)-induced IGF-I expression at tissue level and contributes to GH resistance caused by sepsis/endotoxemia and inflammation. Although the loss of GH responsiveness can be mediated by a
Shirley C Mills et al.
Cellular signalling, 28(4), 316-324 (2016-01-26)
Chemotaxis or directed cell migration is mediated by signalling events initiated by binding of chemokines to their cognate receptors and the activation of a complex signalling cascade. The molecular signalling pathways involved in cell migration are important to understand cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service