Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML0452

Sigma-Aldrich

AZ513

≥98% (HPLC)

Synonym(s):

2,3-Dichloro-N-(1H-pyrrolo[2,3-b]pyridin-5-yl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9Cl2N3O
CAS Number:
Molecular Weight:
306.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

Clc1cccc(C(=O)Nc2cnc3[nH]ccc3c2)c1Cl

InChI

1S/C14H9Cl2N3O/c15-11-3-1-2-10(12(11)16)14(20)19-9-6-8-4-5-17-13(8)18-7-9/h1-7H,(H,17,18)(H,19,20)

InChI key

SIMLBFHHXAMGGM-UHFFFAOYSA-N

Biochem/physiol Actions

AZ513 is a reversible, noncompetitive FAAH inhibitor. It also binds non-competitively at a site distinct from the catalytic site. AZ513 is 20-fold more potent against rat FAAH (IC50 27 nM) than the human form (IC50 551 nM).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service