Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0260

Sigma-Aldrich

ST1936

≥98% (HPLC)

Synonym(s):

2-(5-chloro-2-methyl-1H-indol-3-yl)-N,N-dimethylethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17ClN2
CAS Number:
Molecular Weight:
236.74
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥5 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

CN(C)CCc1c(C)[nH]c2ccc(Cl)cc12

InChI

1S/C13H17ClN2/c1-9-11(6-7-16(2)3)12-8-10(14)4-5-13(12)15-9/h4-5,8,15H,6-7H2,1-3H3

InChI key

KSYMELKKLOFABL-UHFFFAOYSA-N

Biochem/physiol Actions

ST1936 is a high affinity 5-HT6-specific agonist (Ki = 13 nM). The IC50 for competition of LSD from 5-HT6 expressed in HEK cells is 40 nM, and the EC50 for stimulating cAMP production is 16 nM.
ST1936 is a serotonin 5-HT6 agonist

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Srijit Das et al.
eLife, 9 (2020-04-24)
Germ cells are vulnerable to stress. Therefore, how organisms protect their future progeny from damage in a fluctuating environment is a fundamental question in biology. We show that in Caenorhabditis elegans, serotonin released by maternal neurons during stress ensures the

Articles

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service