Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0195

Sigma-Aldrich

Mifamurtide

≥98% (HPLC)

Synonym(s):

CGP-19835, MTP-PE, MTP-cephalin, Muramyl tripeptide phosphatidylethanolamine, N-(N-Acetylmuramoyl)-L-alanyl-D-α-glutaminyl-N-[(7R)-4-hydroxy-4-oxido-10-oxo-7-[(1-oxohexadecyl)oxy]-3,5,9-trioxa-4-phosphapentacos-1-yl]-L-alaninamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H109N6O19P
CAS Number:
Molecular Weight:
1237.50
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
To inquire about this product SML0195, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

water: 2 mg/mL, clear (warmed)

originator

Novartis

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)OCCNC(=O)[C@H](C)NC(=O)CC[C@@H](NC(=O)[C@H](C)NC(=O)[C@@H](C)O[C@@H]([C@H](O)[C@H](O)CO)[C@@H](NC(C)=O)C=O)C(N)=O)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C59H109N6O19P/c1-7-9-11-13-15-17-19-21-23-25-27-29-31-33-52(71)80-41-47(84-53(72)34-32-30-28-26-24-22-20-18-16-14-12-10-8-2)42-82-85(78,79)81-38-37-61-57(75)43(3)62-51(70)36-35-48(56(60)74)65-58(76)44(4)63-59(77)45(5)83-55(54(73)50(69)40-67)49(39-66)64-46(6)68/h39,43-45,47-50,54-55,67,69,73H,7-38,40-42H2,1-6H3,(H2,60,74)(H,61,75)(H,62,70)(H,63,77)(H,64,68)(H,65,76)(H,78,79)/t43-,44-,45+,47+,48+,49-,50+,54+,55+/m0/s1

InChI key

ZVLWUMPAHCEZAW-KRNLDFAISA-N

Application

Mifamurtide may be used in immunological and cancer-related cell signaling studies.

Biochem/physiol Actions

Mifamurtide (Muramyl tripeptide phosphatidylethanolamine; MTP-cephalin; MTP-PE) is a synthetic, lipophilic analog of muramyl dipeptide. It acts as an immunostimulant promoting cancer cell destruction via in vivo macrophage activation.
Mifamurtide is an immunomodulator and regulates the activation of monocytes and macrophages. Mifamurtide upregulates the secretion of pro-inflammatory cytokines such as TNF-α, IL-1, IL-8, nitric oxide and prostaglandins E2 and D2. It has anti-tumor effects in children and young adults with high-grade osteosarcoma.[1][2]
Synthetic, lipophilic analog of muramyl dipeptide. Immunostimulant promoting cancer cell destruction via in vivo macrophage activation. Antineoplastic immunomodulator.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prescrire international, 20(115), 89-89 (2011-06-09)
The standard treatment for children and young adults with osteosarcoma consists of surgery, preceded and followed by methotrexate-based chemotherapy. Mifamurtide is an immunostimulant derived from a bacterial cell wall component. It is authorised in the European Union as an adjunct
L L Worth et al.
Hematology/oncology clinics of North America, 15(4), 723-740 (2001-10-26)
Biologic response modifiers are becoming an important addition to surgery, chemotherapy, and radiotherapy in the management of cancer. As this field of research grows and expands, more biologic response modifiers will be incorporated into therapeutic regimens. By stimulating the immune
Kanji Mori et al.
Expert review of anticancer therapy, 8(2), 151-159 (2008-02-19)
Osteosarcoma is the most common form of primary malignant bone tumor. The use of chemotherapy drugs with many side effects, including high-dose methotrexate, doxorubicin, cisplatin and ifosfamide, has greatly improved osteosarcoma survival compared with surgery alone. However, for 20 years
P Fedorocko et al.
Neoplasma, 50(3), 176-184 (2003-08-26)
The anti-tumor effects of i.p. administered cyclooxygenase inhibitor - diclofenac and i.v. administered liposomal muramyl tripeptide phosphatidylethanolamine (MTP-PE) were investigated using a s.c. growing murine fibrosarcoma tumor. Tumor growth was assessed by measuring tumor volumes and survival of the mice.
James E Frampton
Paediatric drugs, 12(3), 141-153 (2010-05-21)
Mifamurtide (liposomal muramyl tripeptide phosphatidyl ethanolamine; Mepact) is an immunomodulator with antitumor effects that appear to be mediated via activation of monocytes and macrophages. In the EU, mifamurtide is indicated in children, adolescents, and young adults for the treatment of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service